ENA369383653 Enamine

4-bromo-1-(2-methoxyethyl)-3,5-dimethyl-1H-pyrazole

  • MDL number MFCD16687003
  • Cas Number : 1342761-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-99924

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99924
Availability details for
Quantity
  • Availability
  • Add to Cart