ENA369383656 Enamine

[(4-chlorophenyl)(4-fluorophenyl)methyl](methyl)amine

  • MDL number MFCD12472657
  • Cas Number : 1156265-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-99927

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99927
Availability details for
Quantity
  • Availability
  • Add to Cart