ENA369383657 Enamine

2-[1-(2,2,2-trifluoroethyl)piperidin-4-yl]ethan-1-amine

  • MDL number MFCD16048095
  • Cas Number : 1248157-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99928

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99928
Availability details for
Quantity
  • Availability
  • Add to Cart