ENA369383658 Enamine

[6-(4-chlorophenoxy)pyridin-3-yl]methanol

  • MDL number MFCD12403522
  • Cas Number : 1160430-78-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99929

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99929
Availability details for
Quantity
  • Availability
  • Add to Cart