ENA369383667 Enamine

(2-aminoethyl)[(4-fluorophenyl)methyl]methylamine

  • MDL number MFCD12167417
  • Cas Number : 933702-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99811

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99811
Availability details for
Quantity
  • Availability
  • Add to Cart