ENA369383689 Enamine

5-(4-chlorophenyl)-1,2,4-oxadiazol-3-amine

  • MDL number MFCD11109156
  • Cas Number : 23275-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 255...257
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99986

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99986
Availability details for
Quantity
  • Availability
  • Add to Cart