ENA369383694 Enamine

2-hydroxy-3-methyl-2-phenylbutanoic acid

  • MDL number MFCD00021679
  • Cas Number : 15879-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148...150
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99992

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99992
Availability details for
Quantity
  • Availability
  • Add to Cart