ENA370562706 Enamine

5-iodo-N-methylpyrimidin-2-amine

  • MDL number MFCD22369795
  • Cas Number : 16341-50-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101190

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101190
Availability details for
Quantity
  • Availability
  • Add to Cart