ENA370562710 Enamine

octahydro-1H-pyrano[4,3-b]pyridine hydrochloride

  • MDL number MFCD22369801
  • Cas Number : 1384430-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102280

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102280
Availability details for
Quantity
  • Availability
  • Add to Cart