ENA370562719 Enamine

5-acetyl-3-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD22369810
  • Cas Number : 858002-18-3
  • Molecular Weight : 168.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-103004

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103004
Availability details for
Quantity
  • Availability
  • Add to Cart