ENA370562720 Enamine

2-[(2-aminoethyl)sulfanyl]acetic acid hydrochloride

  • MDL number MFCD22369813
  • Cas Number : 60116-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-103254

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103254
Availability details for
Quantity
  • Availability
  • Add to Cart