ENA370562723 Enamine

hex-5-ene-1,2-diamine dihydrochloride

  • MDL number MFCD22369819
  • Cas Number : 1384428-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103676

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103676
Availability details for
Quantity
  • Availability
  • Add to Cart