ENA370562730 Enamine

5-[(dimethylamino)methyl]pyridin-2-amine dihydrochloride

  • MDL number MFCD22369828
  • Cas Number : 1384429-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104190

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-104190
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart