ENA370562731 Enamine

4-(1,2,4-oxadiazol-5-yl)cyclohexan-1-amine hydrochloride

  • MDL number MFCD22369829
  • Cas Number : 1384430-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104256

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104256
Availability details for
Quantity
  • Availability
  • Add to Cart