ENA370562738 Enamine

2-(thiomorpholin-3-yl)ethan-1-ol

  • MDL number MFCD22369836
  • Cas Number : 171351-31-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104304

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104304
Availability details for
Quantity
  • Availability
  • Add to Cart