ENA370562739 Enamine

2-(propan-2-yl)-5H,6H,7H,8H-[1,2,4]triazolo[1,5-a]pyridin-8-amine

  • MDL number MFCD22369837
  • Cas Number : 1384429-13-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104360

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104360
Availability details for
Quantity
  • Availability
  • Add to Cart