ENA370562745 Enamine

1-(2-methoxyphenyl)-2,2-dimethylpropan-1-amine hydrochloride

  • MDL number MFCD22369843
  • Cas Number : 1384431-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 211...213
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104387

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104387
Availability details for
Quantity
  • Availability
  • Add to Cart