ENA370562749 Enamine

[1-(4-methoxyphenyl)-1H-1,2,3-triazol-4-yl]methanamine hydrochloride

  • MDL number MFCD22369847
  • Cas Number : 1384430-87-8
  • Molecular Weight : 240.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [1-(4-methoxyphenyl)-1H-1,2,3-triazol-4-yl]methanamine hydrochloride
Inchi Code 1S/C10H12N4O.ClH/c1-15-10-4-2-9(3-5-10)14-7-8(6-11)12-13-14;/h2-5,7H,6,11H2,1H3;1H
Inchi Key WJUPTUHEDPURIV-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-104414

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-104414
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart