ENA370562751 Enamine

(1R)-1-[5-chloro-2-(difluoromethoxy)phenyl]ethan-1-amine hydrochloride

  • MDL number MFCD22369848
  • Cas Number : 1384435-42-0
  • Molecular Weight : 258.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1R)-1-[5-chloro-2-(difluoromethoxy)phenyl]ethanamine hydrochloride
Inchi Code 1S/C9H10ClF2NO.ClH/c1-5(13)7-4-6(10)2-3-8(7)14-9(11)12;/h2-5,9H,13H2,1H3;1H/t5-;/m1./s1
Inchi Key ZCPBUTRVGCQISG-NUBCRITNSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-104416

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104416
Availability details for
Quantity
  • Availability
  • Add to Cart