ENA370562752 Enamine

(1S)-1-[5-chloro-2-(difluoromethoxy)phenyl]ethan-1-amine hydrochloride

  • MDL number MFCD22369849
  • Cas Number : 1384435-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104417

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104417
Availability details for
Quantity
  • Availability
  • Add to Cart