ENA370562754 Enamine

(1R)-1-[2-(difluoromethoxy)-4-fluorophenyl]ethan-1-amine hydrochloride

  • MDL number MFCD22369851
  • Cas Number : 1384435-41-9
  • Molecular Weight : 241.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1R)-1-[2-(difluoromethoxy)-4-fluorophenyl]ethanamine hydrochloride
Inchi Code 1S/C9H10F3NO.ClH/c1-5(13)7-3-2-6(10)4-8(7)14-9(11)12;/h2-5,9H,13H2,1H3;1H/t5-;/m1./s1
Inchi Key HWFGIYDMPZNFEN-NUBCRITNSA-N

Description

Purity 95
Salt Data Cl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-104419

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104419
Availability details for
Quantity
  • Availability
  • Add to Cart