ENA370562755 Enamine

2-(1-methyl-1H-1,2,4-triazol-5-yl)ethan-1-amine hydrochloride

  • MDL number MFCD22369852
  • Cas Number : 1384427-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104422

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104422
Availability details for
Quantity
  • Availability
  • Add to Cart