ENA370562758 Enamine

2-amino-2-(oxolan-3-yl)ethan-1-ol hydrochloride

  • MDL number MFCD22369855
  • Cas Number : 1384429-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104493

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104493
Availability details for
Quantity
  • Availability
  • Add to Cart