ENA370562765 Enamine

1-cyclopropyl-4-methylpyrrolidin-3-one

  • MDL number MFCD22369862
  • Cas Number : 1384430-91-4
  • Molecular Weight : 139.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-cyclopropyl-4-methyl-3-pyrrolidinone
Inchi Code 1S/C8H13NO/c1-6-4-9(5-8(6)10)7-2-3-7/h6-7H,2-5H2,1H3
Inchi Key NZQACTFKFSQQDY-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-104698

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104698
Availability details for
Quantity
  • Availability
  • Add to Cart