ENA370562768 Enamine

4-cyano-2,6-difluorobenzene-1-sulfonyl chloride

  • MDL number MFCD22369865
  • Cas Number : 1384429-18-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104806

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104806
Availability details for
Quantity
  • Availability
  • Add to Cart