ENA370562772 Enamine

3-ethoxy-4-methylpent-4-enoic acid

  • MDL number MFCD22369869
  • Cas Number : 1384430-96-9
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-ethoxy-4-methyl-4-pentenoic acid
Inchi Code 1S/C8H14O3/c1-4-11-7(6(2)3)5-8(9)10/h7H,2,4-5H2,1,3H3,(H,9,10)
Inchi Key QUCGYSXIZNECFI-UHFFFAOYSA-N

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-104864

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104864
Availability details for
Quantity
  • Availability
  • Add to Cart