ENA370562774 Enamine

3-(dimethylamino)-4-methylbenzene-1-sulfonyl chloride hydrochloride

  • MDL number MFCD22369872
  • Cas Number : 1384431-06-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104998

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104998
Availability details for
Quantity
  • Availability
  • Add to Cart