ENA370562779 Enamine

5-amino-3-methyl-1,2-oxazole-4-sulfonamide

  • MDL number MFCD22369877
  • Cas Number : 1384431-08-6
  • Molecular Weight : 177.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-amino-3-methyl-4-isoxazolesulfonamide
Inchi Code 1S/C4H7N3O3S/c1-2-3(11(6,8)9)4(5)10-7-2/h5H2,1H3,(H2,6,8,9)
Inchi Key GQEVFINHSITALA-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-105126

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105126
Availability details for
Quantity
  • Availability
  • Add to Cart