ENA370562792 Enamine

2,2,2-trifluoro-1-(furan-2-yl)ethan-1-amine hydrochloride

  • MDL number MFCD22369890
  • Cas Number : 65686-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105219

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105219
Availability details for
Quantity
  • Availability
  • Add to Cart