ENA370562796 Enamine

5,7-dichloro-1-methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride

  • MDL number MFCD22369895
  • Cas Number : 1384429-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 284...286
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105291

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105291
Availability details for
Quantity
  • Availability
  • Add to Cart