ENA370562797 Enamine

3-(diethylamino)-2-fluoropropanoic acid hydrochloride

  • MDL number MFCD22369896
  • Cas Number : 1384428-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105293

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105293
Availability details for
Quantity
  • Availability
  • Add to Cart