ENA370562809 Enamine

1-(oxolan-2-yl)-2-(piperazin-1-yl)ethan-1-ol dihydrochloride

  • MDL number MFCD22369910
  • Cas Number : 1384427-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105431

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105431
Availability details for
Quantity
  • Availability
  • Add to Cart