ENA370562813 Enamine

1-(2-bromoethyl)-4-methylpiperidine hydrobromide

  • MDL number MFCD22369914
  • Cas Number : 1384429-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189...191
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105502

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105502
Availability details for
Quantity
  • Availability
  • Add to Cart