ENA370562816 Enamine

4-amino-N-[1-(4-ethoxyphenyl)ethyl]butanamide hydrochloride

  • MDL number MFCD22369917
  • Cas Number : 1384428-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143...145
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105507

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105507
Availability details for
Quantity
  • Availability
  • Add to Cart