ENA370562820 Enamine

2-(2-amino-3,3-dimethylbutoxy)-1,3-dimethylbenzene hydrochloride

  • MDL number MFCD22369921
  • Cas Number : 1384429-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 208...210
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105551

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105551
Availability details for
Quantity
  • Availability
  • Add to Cart