ENA370562827 Enamine

4-[(2-fluorophenyl)methylidene]piperidine hydrochloride

  • MDL number MFCD22369928
  • Cas Number : 183950-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190...192
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105651

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105651
Availability details for
Quantity
  • Availability
  • Add to Cart