ENA370562831 Enamine

1-[(benzyloxy)carbonyl]-3-(carboxymethyl)pyrrolidine-3-carboxylic acid

  • MDL number MFCD22369932
  • Cas Number : 1384430-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105656

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105656
Availability details for
Quantity
  • Availability
  • Add to Cart