ENA370562838 Enamine

3-(3-methylpiperazin-1-yl)pyrazine-2-carbonitrile hydrochloride

  • MDL number MFCD22369940
  • Cas Number : 1384428-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 259...261
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105722

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105722
Availability details for
Quantity
  • Availability
  • Add to Cart