ENA370562843 Enamine

2-[2-(trifluoromethoxy)phenyl]ethan-1-amine hydrochloride

  • MDL number MFCD22369945
  • Cas Number : 1384428-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105823

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105823
Availability details for
Quantity
  • Availability
  • Add to Cart