ENA370562844 Enamine

2-[(3,4-dimethoxyphenyl)methyl]pyrrolidine hydrochloride

  • MDL number MFCD22369946
  • Cas Number : 1384430-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105828

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105828
Availability details for
Quantity
  • Availability
  • Add to Cart