ENA370562848 Enamine

(2-aminopropyl)(methyl)amine dihydrochloride

  • MDL number MFCD22369950
  • Cas Number : 878773-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174...176
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105851

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105851
Availability details for
Quantity
  • Availability
  • Add to Cart