ENA370562851 Enamine

methyl[(1-methyl-1H-1,2,3,4-tetrazol-5-yl)methyl]amine hydrochloride

  • MDL number MFCD22369952
  • Cas Number : 1384429-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105863

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105863
Availability details for
Quantity
  • Availability
  • Add to Cart