ENA370562858 Enamine

N-[2-(4-fluorophenyl)ethyl]cyclopentanamine hydrochloride

  • MDL number MFCD22369960
  • Cas Number : 1384428-50-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-105944

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105944
Availability details for
Quantity
  • Availability
  • Add to Cart