ENA370562859 Enamine

N-[2-(3-fluorophenyl)ethyl]cyclopentanamine hydrochloride

  • MDL number MFCD22369961
  • Cas Number : 1384428-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-105945

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105945
Availability details for
Quantity
  • Availability
  • Add to Cart