ENA370562869 Enamine

5-(aminomethyl)-2,3-dihydro-1lambda6-benzothiophene-1,1-dione hydrochloride

  • MDL number MFCD22369970
  • Cas Number : 1384429-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106026

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106026
Availability details for
Quantity
  • Availability
  • Add to Cart