ENA370562872 Enamine

2-[(acetylsulfanyl)methyl]butanedioic acid

  • MDL number MFCD22369973
  • Cas Number : 192879-20-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106054

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106054
Availability details for
Quantity
  • Availability
  • Add to Cart