ENA370562875 Enamine

2-(aminomethyl)-3-fluoro-N-methylbenzene-1-sulfonamide hydrochloride

  • MDL number MFCD22369977
  • Cas Number : 1384430-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106078

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106078
Availability details for
Quantity
  • Availability
  • Add to Cart