ENA370562877 Enamine

(1R,5S,6R)-3-benzyl-3-azabicyclo[3.1.0]hexan-6-amine dihydrochloride

  • MDL number MFCD22369978
  • Cas Number : 2103395-47-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106208

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106208
Availability details for
Quantity
  • Availability
  • Add to Cart