ENA370562880 Enamine

tert-butyl N-(2-aminohexyl)carbamate

  • MDL number MFCD22369981
  • Cas Number : 1384430-32-3
  • Molecular Weight : 216.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 2-aminohexylcarbamate
Inchi Code 1S/C11H24N2O2/c1-5-6-7-9(12)8-13-10(14)15-11(2,3)4/h9H,5-8,12H2,1-4H3,(H,13,14)
Inchi Key HDYLLORAJORZCO-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-106287

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106287
Availability details for
Quantity
  • Availability
  • Add to Cart