ENA370562894 Enamine

6-bromo-4-methyl-2-oxo-2H-chromene-3-carboxylic acid

  • MDL number MFCD06208402
  • Cas Number : 773109-55-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-104299

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104299
Availability details for
Quantity
  • Availability
  • Add to Cart